Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
Associates with the Ig-mu chain to form a molecular complex that is expressed on the surface of pre-B-cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N27C7-2; pre-B lymphocyte gene 3; Pre-B lymphocyte protein 3; Protein VPreB3
Gene Aliases: 8HS20; N27C7-2; UNQ355/PRO619; VPREB3
UniProt ID: (Human) Q9UKI3
Entrez Gene ID: (Human) 29802
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support