Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAMSFEWPWQ YRFPPFFTLQ PNVDTRQKQL AAWCSLVLSF CRLHKQSSMT VMEAQESPLF NNVKLQRKLP VESIQIVLEE LRKKGNLEWL DKSKSSFLIM WRRPEEWGKL IYQWVSRSGQ NNSVFTLYEL TNGEDTEDEE FHGLDEATLL RALQALQQEH KAEIITVSDG RGVKFF
VPS25 (Vacuolar Protein Sorting 25 Homolog) is a Protein Coding gene. Among its related pathways are Delta508-CFTR traffic / ER-to-Golgi in CF and Budding and maturation of HIV virion. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and protein N-terminus binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dermal papilla-derived protein 9; ELL-associated protein of 20 kDa; ESCRT-II complex subunit VPS25; HGNC:28122; hVps25; MGC10540; Vacuolar protein-sorting-associated protein 25
Gene Aliases: DERP9; EAP20; FAP20; VPS25
UniProt ID: (Human) Q9BRG1
Entrez Gene ID: (Rabbit) 103345024, (Human) 84313
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support