Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases:
FLJ12930; H beta 58; H
Gene Aliases: AA407240; HB58; Hbeta58; PEP8A; VPS26; VPS26A
UniProt ID: (Human) O75436, (Rat) Q6AY86, (Mouse) P40336
Entrez Gene ID: (Human) 9559, (Rat) 361846, (Mouse) 30930
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support