Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is rat liver tissue lysate.
PA5-21104 can be used with blocking peptide PEP-1218.
VPS39 is the mammalian homolog of the yeast vacuolar protein VAM6, that in mammals, promotes lysosome clustering and fusion in vivo. VPS39 is also thought to modulate the TGF-beta response by coupling the TGF-beta receptor complex to the Smad pathway. Similar to the related protein TRAP1, the VPS39 protein is essential for early embryonic development, with mice lacking VPS39 dying at or before E6. 5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hVam6p; OTTMUSP00000023341; OTTMUSP00000023350; PI3K type 3; TRAP1-like protein; vacuolar protein sorting 39 homolog; Vam6; Vam6/Vps39-like protein; VPS39 HOPS complex subunit
Gene Aliases: A230065P22Rik; AW743070; hVam6p; KIAA0770; mVam6; Pldn; TLP; VAM6; Vam6P; VPS39
UniProt ID: (Human) Q96JC1, (Mouse) Q922I3
Entrez Gene ID: (Human) 23339, (Rat) 362199, (Mouse) 269338
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support