Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FYANNMYLN FAEIGSNIKN LMEDFQKKKP KEQQKLESIA DMKAFVENYP QFKKMSGTVS KHVTVVGELS RLVSERNLLE VSEVEQELAC QNDHSSALQN IKRLLQNPKV TEFDAARLVM LYALHYERHS SNSLPGLMMD LRNKGVSEKY RKLVSAVVEY GGKRVRGSDL FSPKDAVAIT KQFLKGLKGV ENVYTQHQPF LHETLDHLIK GRLKENLYPY LGPSTLRDRP QDIIVFVIGG ATYEEALTVY NLNRTTPGVR IVLGGTTVHN TKSFLEEVLA SGLHSRSKES SQVTSRSASR R (271-570 aa encoded by BC028382)
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting genes involved in vesicle transport to vacuoles. This gene is a member of the Sec1 domain family, and shows a high degree of sequence similarity to mouse, rat and yeast Vps45. The exact function of this gene is not known, but its high expression in peripheral blood mononuclear cells suggests a role in trafficking proteins, including inflammatory mediators.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: h-VPS45; HGNC:14579; hlVps45; leucocyte vacuolar protein sorting 45; mVps45; RP11-458I7.2; rVps45; vacuolar protein sorting 45A; vacuolar protein sorting protein 45; Vacuolar protein sorting-associated protein 45; vesicular transport protein rvps45
Gene Aliases: AI462172; AW554165; H1; H1VPS45; mVps45; SCN5; VPS45; VPS45A; VPS45B; VPS54A; VSP45; VSP45A
UniProt ID: (Human) F5H8K1, (Mouse) P97390, (Rat) O08700
Entrez Gene ID: (Human) 11311, (Mouse) 22365, (Rat) 64516
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support