Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KENQSEGKG SDSDSEGDNP EKKKLQEQLM GAVVMEKPNI RWNDVAGLEG AKEALKEAVI LPIKFPHLFT GKRTPWRGIL LFGPPGTGKS YLAKAVATEA NNSTFFSVSS SDLMSKWLGE SEKLVKNLFE LARQHKPSII FIDEVDSLCG SRNENESEAA RRIKTEFLVQ MQGVGNNNDG TLVLGATNIP WVLDSAIRRR FEKRIYIPLP EEAARAQMFR LHLGSTPHNL TDANIHELAR KTEGYSGADI SIIVRDSLMQ PVRKVQSATH FKKVCGPSRT NPSMMIDDLL TPCSPGDPGA MEMTWMDVPG DKLLEPVVCM SDMLRSLATT RPTVNADDLL KVKKFSEDFG QES (86-437 aa encoded by BC047932)
The protein encoded by this gene is a member of the AAA protein family (ATPases associated with diverse cellular activities), and is the homolog of the yeast Vps4 protein. In humans, two paralogs of the yeast protein have been identified. The former share a high degree of aa sequence similarity with each other, and also with yeast Vps4 and mouse Skd1 proteins. The mouse Skd1 (suppressor of K+ transport defect 1) has been shown to be really an yeast Vps4 ortholog. Functional studies indicate that both human paralogs associate with the endosomal compartments, and are involved in intracellular protein trafficking, similar to Vps4 protein in yeast. The gene encoding this paralog has been mapped to chromosome 16; the gene for the other resides on chromosome 18.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hVPS4; Protein SKD2; SKD1-homolog; vacuolar protein sorting 4a; vacuolar protein sorting factor 4A; vacuolar protein sorting protein 4a; Vacuolar protein sorting-associated protein 4A; vacuolar sorting protein 4; VPS4-1; VPS4A
Gene Aliases: 4930589C15Rik; AI325971; AW553189; SKD1; SKD1A; SKD2; VPS4; VPS4-1; vps4-A; VPS4A
UniProt ID: (Human) Q9UN37, (Rat) Q793F9, (Mouse) Q8VEJ9
Entrez Gene ID: (Human) 27183, (Rat) 246772, (Mouse) 116733
Molecular Function:
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support