Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
This gene encodes for a protein that in yeast forms part of a trimeric vacuolar-protein-sorting complex that is required for retrograde transport of proteins from prevacuoles to the late Golgi compartment. As in yeast, mammalian Vps54 proteins contain a coiled-coil region and dileucine motifs. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hepatocellular carcinoma protein 8; protein phosphatase 1, regulatory subunit 164; Tumor antigen HOM-HCC-8; Tumor antigen SLP-8p; Tumor antigen SLP-8p homolog; vacuolar protein sorting 54 homolog; Vacuolar protein sorting-associated protein 54; VPS54 GARP complex subunit; VPS54, GARP complex subunit; Vps54-like protein
Gene Aliases: HCC8; hVps54L; mSLP8; PPP1R164; SLP-8p; VPS54; VPS54L; Vsp54; WR
UniProt ID: (Human) Q9P1Q0, (Rat) Q9JMK8, (Mouse) Q5SPW0
Entrez Gene ID: (Human) 51542, (Pig) 100621212, (Rat) 286932, (Mouse) 245944
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support