Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
VRK2 serine/threonine kinases vaccinia related kinase 2 is a member of the vaccinia-related kinase (VRK) family of kinases. VRK2 is widely expressed in human tissues and highly expressed in actively dividing cells, such as those in testis, leukocytes, fetal liver and carcinomas. VRK2 can be used to phosphorylate casein and itself undergoes autophosphorylation. VRK2 interacts specifically with Epstein-Barr virus BHRF1, a homologue of Bcl-2, and this interaction is involved in protecting cells from apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Serine/threonine-protein kinase VRK2; vaccinia virus B1R-related kinase 2; vaccinia virus BIR kinase related kinase 2; Vaccinia-related kinase 2
Gene Aliases: 2810003O05Rik; AI447698; VRK2
UniProt ID: (Human) Q86Y07, (Mouse) Q8BN21
Entrez Gene ID: (Human) 7444, (Mouse) 69922
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support