Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FARNKAKAK AKERNSKTIA ELEPMTKINP RGESEAMPRE DATQLEVTLP SSIHETGPDT IQEPDYEPKP TQEPAPEPAP GSEPMAVPDL DIELELEPET QSELEPEPEP EPESEPGVVV EPLSEDEKGV VKA (256-387 aa encoded by BC043216)
VSIG1 encodes a member of the junctional adhesion molecule (JAM) family. The encoded protein contains multiple glycosylation sites at the N-terminal region, and multiple phosphorylation sites and glutamic acid/proline (EP) repeats at the C-terminal region. The gene is expressed in normal stomach and testis, as well as in gastric, esophageal and ovarian cancers. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell surface A33 antigen; Glycoprotein A34; MGC44287; V-set and immunoglobulin domain-containing protein 1
Gene Aliases: 1700062D20Rik; dJ889N15.1; GPA34; VSIG1
UniProt ID: (Human) Q86XK7
Entrez Gene ID: (Human) 340547
Molecular Function: defense/immunity protein immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support