Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
C6ORF55 encodes a protein involved in trafficking of the multivesicular body, an endosomal compartment involved in sorting membrane proteins for degradation in lysosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dopamine-responsive gene 1 protein; DRG-1; homolog of mouse SKD1-binding protein 1; LIP5; LYST-interacting protein 5; SBP1; SKD1-binding protein 1; Vacuolar protein sorting-associated protein VTA1 homolog; vesicle (multivesicular body) trafficking 1; Vps20-associated 1 homolog
Gene Aliases: C6orf55; DRG-1; DRG1; HSPC228; LIP5; My012; SBP1; VTA1
UniProt ID: (Human) Q9NP79
Entrez Gene ID: (Human) 51534
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support