Search Thermo Fisher Scientific
HYB 060-01-02 is specific for human vWf from serum or plasma and has been successfully used in ELISA procedures. HYB 060-01-02 epitope specificity differs from that of HYB 060-02-02.
The HYB 060-01-02 immunogen is Factor VIII complex isolated from human plasma.
NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.