Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ECGTDALRF GLCAYMSQGR DINLDVNRIL GYRHFCNKLW NATKFALRGL GKGFVPSPTS QPGGHESLVD RWIRSRLTEA VRLSNQGFQA YDFPAVTTAQ YSFWLYELCD VYLECLKPVL NGVDQVAAEC ARQTLYTCLD VGLRLLSPFM PFVTEELFQR LPRRMPQAPP SLCVTPYPEP SECSWKDPEA EAALELALSI TRAVRSLRAD YNLTRIRPDC FLEVADEATG ALASAVSGYV QALASAGVVA VLALGAPAPQ GCAVALASDR CSIHLQLQGL VDPARELGKL QAKRVEAQRQ AQRLRERRAA SGYPVKVPLE VQEADEAKLQ QTEAELRKVD EAIALFQKML (916-1264 aa encoded by BC012808)
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. This protein belongs to the class-I aminoacyl-tRNA synthetase family and is located in the class III region of the major histocompatibility complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 6.1.1.9; Protein G7a; valine tRNA ligase 1, cytoplasmic; Valine--tRNA ligase; ValRS; valyl trna synthetase; Valyl-tRNA synthetase; valyl-tRNA synthetase 2
Gene Aliases: Bat6; D17H6S56E; G7A; TrsVal; VARS; VARS1; VARS2
UniProt ID: (Human) Q9UQM2, (Rat) Q04462, (Mouse) Q9Z1Q9
Entrez Gene ID: (Human) 7407, (Rat) 25009, (Mouse) 22321
Molecular Function:
aminoacyl-tRNA synthetase
translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support