Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: RKEKNGWRTH GRNGTENINH RGGYHGGSSR SRSSIFHAGK SQGLHENNIP
Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%
Vasculin is a novel vascular protein differentially expressed in human atherogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP761C169; GC-rich promoter-binding protein 1; Vascular wall-linked protein; Vasculin
Gene Aliases: GPBP; GPBP1; SSH6; VASCULIN
UniProt ID: (Human) Q86WP2
Entrez Gene ID: (Human) 65056
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support