Product References
A FACS-based novel isolation technique identifies heterogeneous CTCs in oral squamous cell carcinoma.
Frontiers in oncology
Chauhan A,Pal A,Sachdeva M,Boora GS,Parsana M,Bakshi J,Verma RK,Srinivasan R,Chatterjee D,Maitra A,Ghoshal S
Published figure using Vimentin monoclonal antibody (Product # 14-9897-82) in Flow Cytometry
Wed Mar 13 00:00:00 UTC 2024
Matrix stiffness induces epithelial-to-mesenchymal transition via Piezo1-regulated calcium flux in prostate cancer cells.
iScience
Lopez-Cavestany M,Hahn SB,Hope JM,Reckhorn NT,Greenlee JD,Schwager SC,VanderBurgh JA,Reinhart-King CA,King MR
Published figure using Vimentin monoclonal antibody (Product # 14-9897-82) in Western Blot
Fri Apr 21 00:00:00 UTC 2023
Transcriptome analyses of the cortex and white matter of focal cortical dysplasia type II: Insights into pathophysiology and tissue characterization.
Frontiers in neurology
Assis-Mendonça GR,Athié MCP,Tamanini JVG,de Souza A,Zanetti GG,Araújo PAORA,Ghizoni E,Tedeschi H,Alvim MKM,de Almeida VS,de Souza W,Coras R,Yasuda CL,Blümcke I,Vieira AS,Cendes F,Lopes-Cendes I,Rogerio F
14-9897-82 was used in Immunohistochemistry to evaluate the transcriptomes of the gray and white matters of surgical focal cortical dysplasia type II specimens.
Tue Apr 04 00:00:00 UTC 2023
Optimizing culturing conditions in patient derived 3D primary slice cultures of head and neck cancer.
Frontiers in oncology
Greier MDC,Runge A,Dudas J,Carpentari L,Schartinger VH,Randhawa A,Mayr M,Petersson M,Riechelmann H
14-9897-82 was used in Immunohistochemistry to optimize cultivation conditions for HNC SC and analyze the added effects of platelet rich fibrin (PRF) on these conditions.
Tue Apr 18 00:00:00 UTC 2023
Aspirin increases the efficacy of gemcitabine in pancreatic cancer by modulating the PI3K/AKT/mTOR signaling pathway and reversing epithelial‑mesenchymal transition.
Oncology letters
Zhou H,Yun X,Shu Y,Xu K
14-9897-82 was used in Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot to reveal that aspirin (ASA) increased the efficacy of gemcitabine in reducing the proliferation, migration and invasion of pancreatic cancer cells and increasing their apoptosis.
Wed Mar 01 00:00:00 UTC 2023
Aspirin increases the efficacy of gemcitabine in pancreatic cancer by modulating the PI3K/AKT/mTOR signaling pathway and reversing epithelial‑mesenchymal transition.
Oncology letters
Zhou H,Yun X,Shu Y,Xu K
14-9897-82 was used in Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot to reveal that aspirin (ASA) increased the efficacy of gemcitabine in reducing the proliferation, migration and invasion of pancreatic cancer cells and increasing their apoptosis.
Wed Mar 01 00:00:00 UTC 2023
Theranostics application of tumor-initiating cell probe TiY in non-small cell lung cancer.
Theranostics
Lee YA,Lek CCJ,Rong G,Wu Z,Shathishwaran S,Lee JHJ,Tam WL,Wuestefeld T,Park SJ,Jung S,Kim B,Kang NY,Chang YT
Published figure using Vimentin monoclonal antibody (Product # 14-9897-82) in Western Blot
Mon Mar 20 00:00:00 UTC 2023
Theranostics application of tumor-initiating cell probe TiY in non-small cell lung cancer.
Theranostics
Lee YA,Lek CCJ,Rong G,Wu Z,Shathishwaran S,Lee JHJ,Tam WL,Wuestefeld T,Park SJ,Jung S,Kim B,Kang NY,Chang YT
Published figure using Vimentin monoclonal antibody (Product # 14-9897-82) in Western Blot
Mon Mar 20 00:00:00 UTC 2023
Human neural cell type-specific extracellular vesicle proteome defines disease-related molecules associated with activated astrocytes in Alzheimer's disease brain.
Journal of extracellular vesicles
You Y,Muraoka S,Jedrychowski MP,Hu J,McQuade AK,Young-Pearse T,Aslebagh R,Shaffer SA,Gygi SP,Blurton-Jones M,Poon WW,Ikezu T
14-9897-82 was used in Immunocytochemistry to identify cell type-specific extracellular vesicle (EV) protein markers in human-induced pluripotent stem cell-derived neural cell types, including excitatory neurons (ATP1A3, NCAM1), astrocytes (LRP1, ITGA6), microglia-like cells (ITGAM, LCP1), and oligodendrocyte-like cells (LAMP2, FTH1), along with 16 pan-EV marker candidates.
Sat Jan 01 00:00:00 UTC 2022
Conjugation of VEGFR1/R2-targeting peptide with gold nanoparticles to enhance antiangiogenic and antitumoral activity.
Journal of nanobiotechnology
Zanjanchi P,Asghari SM,Mohabatkar H,Shourian M,Shafiee Ardestani M
14-9897-82 was used in Western Blot to suggest that GNP-VGB3 is a promising candidate for clinical translation.
Tue Jan 04 00:00:00 UTC 2022
Conjugation of VEGFR1/R2-targeting peptide with gold nanoparticles to enhance antiangiogenic and antitumoral activity.
Journal of nanobiotechnology
Zanjanchi P,Asghari SM,Mohabatkar H,Shourian M,Shafiee Ardestani M
14-9897-82 was used in Western Blot to suggest that GNP-VGB3 is a promising candidate for clinical translation.
Tue Jan 04 00:00:00 UTC 2022
Characterization of epithelial cells, connective tissue cells and immune cells in human upper airway mucosa by immunofluorescence multichannel image cytometry: a pilot study.
Histochemistry and cell biology
Giotakis AI,Dudas J,Glueckert R,Dejaco D,Ingruber J,Fleischer F,Innerhofer V,Pinggera L,Bektic-Tadic L,Gabriel SAM,Riechelmann H
Published figure using Vimentin monoclonal antibody (Product # 14-9897-82) in Immunocytochemistry
Mon Mar 01 00:00:00 UTC 2021
Characterization of epithelial cells, connective tissue cells and immune cells in human upper airway mucosa by immunofluorescence multichannel image cytometry: a pilot study.
Histochemistry and cell biology
Giotakis AI,Dudas J,Glueckert R,Dejaco D,Ingruber J,Fleischer F,Innerhofer V,Pinggera L,Bektic-Tadic L,Gabriel SAM,Riechelmann H
Published figure using Vimentin monoclonal antibody (Product # 14-9897-82) in Immunocytochemistry
Mon Mar 01 00:00:00 UTC 2021
MicroRNA-384 inhibits nasopharyngeal carcinoma growth and metastasis via binding to Smad5 and suppressing the Wnt/β-catenin axis.
Cytotechnology
Zeng X,Liao H,Wang F
14-9897-82 was used in Western Blot to show that miR-384 targets Smad5 and inactivates the Wnt/β-catenin pathway, which exerts a suppressing role in NPC cell behaviors as well as tumor growth in vivo.
Thu Apr 01 00:00:00 UTC 2021
MicroRNA-384 inhibits nasopharyngeal carcinoma growth and metastasis via binding to Smad5 and suppressing the Wnt/β-catenin axis.
Cytotechnology
Zeng X,Liao H,Wang F
14-9897-82 was used in Western Blot to show that miR-384 targets Smad5 and inactivates the Wnt/β-catenin pathway, which exerts a suppressing role in NPC cell behaviors as well as tumor growth in vivo.
Thu Apr 01 00:00:00 UTC 2021
Induction of Epithelial-mesenchymal Transition in MDCK II Cells.
Bio-protocol
Pastuła A,Lundmark R
14-9897-82 was used in Immunohistochemistry to provide a detailed protocol on how to induce EMT in Madin-Darby Canine Kidney (MDCK) II epithelial cell line and perform immunofluorescent staining on EMT-induced cells.
Fri Feb 05 00:00:00 UTC 2021
Nonmuscle Myosin II Activation Regulates Cell Proliferation, Cell Contraction, and Myofibroblast Differentiation in Keloid-Derived Fibroblasts.
Advances in wound care
Lu YY,Fang CC,Hong CH,Wu CH,Lin YH,Chang KL,Lee CH
14-9897 was used in Immunohistochemistry-immunofluorescence to prove NM-IIA might be one of the therapeutic targets in keloids.
Tue Sep 01 00:00:00 UTC 2020
miR-16 enhances miR-302/367-induced reprogramming and tumor suppression in breast cancer cells.
IUBMB life
Hoseinbeyki M,Taha MF,Javeri A
14-9897-82 was used in Western Blot to study the impact of exogenous induction of miR-16 in MDA-MB-231 and SK-BR-3 breast cancer cells following overexpression of miR-302/367 cluster and investigate whether transfection of these cells by a mature miR-16 mimic could affect the reprogramming state of the cells and their tumorigenicity.
Fri May 01 00:00:00 UTC 2020
Omentin-1 is associated with atrial fibrillation in patients with cardiac valve disease.
BMC cardiovascular disorders
Chen Y,Liu F,Han F,Lv L,Tang CE,Xie Z,Luo F
14-9897 was used in Western Blotting to explore the role of omentin-1 in patient samples with atrial fibrillation.
Wed May 06 00:00:00 UTC 2020
Omentin-1 is associated with atrial fibrillation in patients with cardiac valve disease.
BMC cardiovascular disorders
Chen Y,Liu F,Han F,Lv L,Tang CE,Xie Z,Luo F
14-9897 was used in Western Blotting to explore the role of omentin-1 in patient samples with atrial fibrillation.
Wed May 06 00:00:00 UTC 2020
Omentin-1 is associated with atrial fibrillation in patients with cardiac valve disease.
BMC cardiovascular disorders
Chen Y,Liu F,Han F,Lv L,Tang CE,Xie Z,Luo F
14-9897 was used in Western Blotting to explore the role of omentin-1 in patient samples with atrial fibrillation.
Wed May 06 00:00:00 UTC 2020
GMP-compatible and xeno-free cultivation of mesenchymal progenitors derived from human-induced pluripotent stem cells.
Stem cell research & therapy
McGrath M,Tam E,Sladkova M,AlManaie A,Zimmer M,de Peppo GM
Published figure using Vimentin monoclonal antibody (Product # 14-9897-82) in Immunocytochemistry
Fri Jan 11 00:00:00 UTC 2019
Levels of hepatic Th17 cells and regulatory T cells upregulated by hepatic stellate cells in advanced HBV-related liver fibrosis.
Journal of translational medicine
Li X,Su Y,Hua X,Xie C,Liu J,Huang Y,Zhou L,Zhang M,Li X,Gao Z
Published figure using Vimentin monoclonal antibody (Product # 14-9897-82) in Immunofluorescence
Tue Apr 11 00:00:00 UTC 2017
Interaction of TWEAK with Fn14 leads to the progression of fibrotic liver disease by directly modulating hepatic stellate cell proliferation.
The Journal of pathology
Wilhelm A,Shepherd EL,Amatucci A,Munir M,Reynolds G,Humphreys E,Resheq Y,Adams DH,Hübscher S,Burkly LC,Weston CJ,Afford SC
14-9897 was used in Immunofluorescence to conclude that TWEAK/Fn14 interaction leads to the progression of fibrotic liver disease via direct modulation of HSCs proliferation, making it a potential therapeutic target for liver fibrosis.
Sun May 01 00:00:00 UTC 2016
Biphasic Squamoid Alveolar Renal Cell Carcinoma: A Distinctive Subtype of Papillary Renal Cell Carcinoma?
The American journal of surgical pathology
Hes O,Condom Mundo E,Peckova K,Lopez JI,Martinek P,Vanecek T,Falconieri G,Agaimy A,Davidson W,Petersson F,Bulimbasic S,Damjanov I,Jimeno M,Ulamec M,Podhola M,Sperga M,Pane Foix M,Shelekhova K,Kalusova K,Hora M,Rotterova P,Daum O,Pivovarcikova K,Michal M
14989782 was used in immunohistochemistry - paraffin section to characterize 21 cases of biphasic squamoid alveolar renal cell carcinoma
Sun May 01 00:00:00 UTC 2016
Expression of intercellular adhesion molecule 1 by hepatocellular carcinoma stem cells and circulating tumor cells.
Gastroenterology
Liu S,Li N,Yu X,Xiao X,Cheng K,Hu J,Wang J,Zhang D,Cheng S,Liu S
14-9897 was used in Flow cytometry/Cell sorting to demonstrate that the stem cell factor Nanog regulates expression of ICAM-1 in hepatocellular carcinoma cells.
Wed May 01 00:00:00 UTC 2013