Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: PC-3, Mouse spleen, Rat lung, Rat spleen, Rat kidney
Immunogen sequence: EKINFSGGEP FLQDRGEYLG KLVRFCKVEL RLPSVSIVSN GSLIRERWFQ NYGEYLDILA ISCDSFDEEV NVLIGRGQGK KNHVENLQKL RRWCRDYRVA FKINSVINRF NVEEDMTEQI KALNPVRWKV FQCLLIEGEN CGEDALREAE RFVIGDEEFE RFLERHKEVS CLVPESNQKM KDSYLILDEY MRFLNCRKGR KDPSKSILDV GVEEAIKFSG FDEKMFLKRG GKYIWSKADL KLDW
Viperin is a 43kDa protein which is one of several hundred interferon inducible viral proteins. Elevated expression of viperin is induced by LPS and poly I:C through TLR dependent signaling pathways. Sendai virus mediated viperin expression is induced independently of TLR signaling pathways through RLRs - RIG-I and MAVs (Severa, 2006). Viperin has been shown to play an important role in interferon mediated anti-viral activity in a Hepatitis C virus model system (Helbig, 2011). Viperin is induced in lymphoid cells and dendritic cells (DCs) and highly induced in neutrophils and macrophages during acute LCMV infection in mice (Hinson, ). Viperin functions as a mediator of viral resistance and as a marker for interferon responsiveness making its detection and mechanism of action through specific antibody probes valuable for further studies.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bone-expressed sequence tag 5 protein; Cytomegalovirus-induced gene 5 protein; Radical S-adenosyl methionine domain-containing protein 2; RSAD2; rsad2 {ECO:0000250|UniProtKB:Q8WXG1}; S-adenosylmethionine-dependent nucleotide dehydratase RSAD2; SAND; VHSV-induced-like protein; Viperin; viral hemorrhagic septicemia virus (VHSV)-induced gene 1; Virus inhibitory protein, endoplasmic reticulum-associated, interferon-inducible
Gene Aliases: 2510004L01Rik; Best5; cig33; CIG5; RSAD2; Vig1
UniProt ID: (Human) Q8WXG1, (Rat) O70600, (Mouse) Q8CBB9
Entrez Gene ID: (Human) 91543, (Rat) 65190, (Mouse) 58185
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support