Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
A suggested positive control is human testis tissue lysate.
PA5-34536 can be used with blocking peptide PEP-1579.
The WW domain containing adaptor with coiled-coil protein (WAC) contains a WW domain that mediates protein-protein interactions and colocalizes with RNA splicing factor SC35. Further studies have indicated that WAC is a functional partner of the RNF20/40 complex that ubiquitinates Histone H2B, and that WAC regulates H2B ubiquitination. WAC targets RNF20/40 to associate with RNA polymerase II complex for H2B ubiquitination at active transcription sites. WAC-dependent transcription is also important for cell-cycle checkpoint activation in response to genotoxic stress.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: WW domain-containing adapter protein with coiled-coil; WW domain-containing adaptor with coiled-coil; WW domain-containing protein 4
Gene Aliases: 1110067P07Rik; A230035H12Rik; AI256735; AI256776; BM-016; DESSH; KIAA1844; PRO1741; RGD1562407; WAC; Wwp4
UniProt ID: (Human) Q9BTA9, (Mouse) Q924H7
Entrez Gene ID: (Human) 51322, (Rat) 307029, (Mouse) 225131
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support