Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse
Transcriptional repressor which may play a role in development of the central nervous system (CNS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: friend of EBNA2 (Epstein-Barr virus nuclear protein 2); Friend of EBNA2 protein; WAPL cohesin release factor; Wings apart-like protein homolog
Gene Aliases: FOE; KIAA0261; WAPAL; WAPL
UniProt ID: (Human) Q7Z5K2
Entrez Gene ID: (Human) 23063
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support