Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
Positive Control - WB: human Hela whole cell, human 293T whole cell, human HepG2 whole cell, rat L6 whole cell, mouse C2C12 whole cell. IHC: mouse colon tissue, human lymphoma tissue, human stomach cancer tissue, human tonsil tissue. ICC/IF: T-47D cell. Flow: A431 cell.|Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Transcriptional repressor which may play a role in development of the central nervous system (CNS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DIF-2; Dioxin-inducible factor 2; friend of EBNA2; friend of EBNA2 (Epstein-Barr virus nuclear protein 2); Friend of EBNA2 protein; WAPL cohesin release factor; wings apart-like homolog (Drosophila); Wings apart-like protein homolog
Gene Aliases: A530089A20Rik; BC037674; DIF-2; FOE; KIAA0261; RGD1308960; WAPAL; WAPL
UniProt ID: (Human) Q7Z5K2, (Mouse) Q65Z40
Entrez Gene ID: (Human) 23063, (Rat) 290577, (Mouse) 218914
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support