Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
A recommended positive control is A-20 cell lysate.
At least four isoforms of WAPL are known to exist.
Transcriptional repressor which may play a role in development of the central nervous system (CNS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DIF-2; Dioxin-inducible factor 2; friend of EBNA2; friend of EBNA2 (Epstein-Barr virus nuclear protein 2); Friend of EBNA2 protein; WAPL cohesin release factor; wings apart-like homolog (Drosophila); Wings apart-like protein homolog
Gene Aliases: A530089A20Rik; BC037674; DIF-2; FOE; KIAA0261; RGD1308960; WAPAL; WAPL
UniProt ID: (Human) Q7Z5K2, (Mouse) B9EIG8
Entrez Gene ID: (Human) 23063, (Rat) 290577, (Mouse) 218914
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support