Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MALHSMRKAR ERWSFIRALH KGSAAAPALQ KDSKKRVFSG IQPTGILHLG NYLGAIESWV RLQDEYDSVL YSIVDLHSIT VPQDPAVLRQ
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. This gene encodes the mitochondrial tryptophanyl-tRNA synthetase. Two alternative transcripts encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (Mt)TrpRS; TrpRS; tryptophan tRNA ligase 2, mitochondrial; Tryptophan--tRNA ligase, mitochondrial; Tryptophanyl-tRNA synthetase
Gene Aliases: TrpRS; WARS2
UniProt ID: (Human) Q9UGM6
Entrez Gene ID: (Human) 10352
Molecular Function: aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support