Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSVQQQPPP PRRVTNVGSL LLTPQENESL FTFLGKKCVT MSSAVVQLYA ADRNCMWSKK CSGVACLVKD NPQRSYFLRI FDIKDGKLLW EQELYNNFVY NSPRGYFHTF AGDTCQVALN FANEEEAKKF RKAVTDLLGR RQRKSEKRRD PPNGPNLPMA TVDIKNPEIT TNRFYGPQVN NISHTKEKKK GKAKKKRLTK ADIGTPSNFQ HIGHVGWDPN TGFDLNNLDP ELKNLFDMCG ISEAQLKDRE TSKVIYDFIE KTGGVEAVKN ELRRQAPPPP PPSRGGPPPP PPPPHNSGPP PPPARGRGAP PPPPSRAPTA APPPPPPSRP SVAVPPPPPN RMYPPPPPAL PSSAPSGPPP PPPSVLGVGP VAPPPPPPPP PPPGPPPPPG LPSDGDHQVP TTAGNKAALL DQIREGAQLK KVEQNSRPVS CSGRDALLDQ IRQGIQLKSV ADGQESTPPT PAPTSGIVGA LMEVMQKRSK AIHSSDEDED EDDEEDFEDD DEWED
The Wiskott-Aldrich syndrome (WAS) family of proteins share similar domain structure, and are involved in transduction of signals from receptors on the cell surface to the actin cytoskeleton. The presence of a number of different motifs suggests that they are regulated by a number of different stimuli, and interact with multiple proteins. Recent studies have demonstrated that these proteins, directly or indirectly, associate with the small GTPase, Cdc42, known to regulate formation of actin filaments, and the cytoskeletal organizing complex, Arp2/3. The WASL gene product is a homolog of WAS protein, however, unlike the latter, it is ubiquitously expressed and shows highest expression in neural tissues. It has been shown to bind Cdc42 directly, and induce formation of long actin microspikes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin nucleation-promoting factor WASL; DKFZp779G0847; MGC48327; n wasp; N-WASP; Neural Wiskott-Aldrich syndrome protein; Wiskott-Aldrich syndrome-like
Gene Aliases: N-WASP; NWASP; WASL; WASPB
UniProt ID: (Human) O00401
Entrez Gene ID: (Human) 8976
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support