Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GTNVEGVSSH QSAPPHQEGE PGAGVTPAST PPSCRYRRLT GDSGIELCPC PASGEGEPVK EVRVSATLPD LEDYSPCALP PESVPQIFPM GLSSSEGD
The globular WW domain, named for the conserved tryptophan residues in the protein motif present in various structural and regulatory proteins, is known to play a role in the mediation of protein-protein interactions. This gene encodes a ligand of the WW domain of the Yes kinase-associated protein. Readthrough transcription of the neighboring upstream gene, which encodes INO80 complex subunit B, into this gene generates a non-coding transcript.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: WBP-1; WW domain-binding protein 1
Gene Aliases: WBP-1; WBP1
UniProt ID: (Human) Q96G27
Entrez Gene ID: (Human) 23559
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support