Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MATEGGGKEM NEIKTQFTTR EGLYKLLPHS EYSRPNRVPF NSQGSNPVRV SFVNLNDQSG NGDRLCFNVG RELYFYIYKG VRKAADLSKP IDKRIYKGTQ PTCHDFNHLT ATAESVSLLV GFSAGQVQLI DPIKKETSKL FNEERLIDKS RVTCVKWVHG SESLFLVAHS SGNMYLYNVE HTCGTTAPHY QLLKQGESFA VHTCKSKSTR NPLLKWTVGE GALNEFAFSP DGKFLACVSQ DGFLRVFNFD SVELHGTMKS YFGGLLCVCW SPDGKYIVTG GEDDLVTVWS FVDCRVIARG HGHKSWVSVV AFDPYTTSVE EGDPMEFSGS DEDFQDLLHF GRDRANSTQS RLSKRNSTDS RPVSVTYRFG SVGQDTQLCL WDLTEDILFP HQPLSRARTH TNVMNATSPP AGSNGNSVTT PGNSVPPPLP RSNSLPHSAV SNAGSKSSVM DGAIASGVSK FATLSLHDRK ERHHEKDHKR NHSMGHISSK SSDKLNLVTK TKTDPAKTLG TPLCPRMEDV PLLEPLICKK IAHERLTVLI FLEDCIVTAC QEGFICTWGR PGKVVSFNP
CUL-2 is a member of the family of human cullin genes (CUL-1, -2, -3, -4a, -4b, and -5), homologous to the S. cerevisiae cdc53 gene. CUL-1 forms a complex with human p19skp1 and F box protein p45skp2. Together with cdc53 (Known as CulA), as three subunits, they form a ubiquitin ligase controlling proteolysis of G1 cell cycle regulatory proteins. In contrast to CUL-1, CUL-2 does not associate with p19skp1 and p45skp2. Instead, CUL-2 forms a complex with an inactive transcriptional elongation complex, SIII, formed by three subunits: Elongain C, Elongin B and VHL. The function of this association remains unknown.
仅用于科研。不用于诊断过程。未经明确授权不得转售。
蛋白别名: Protein DMR; WD repeat-containing protein 20
基因别名: DMR; WDR20
UniProt ID: (Human) Q8TBZ3
Entrez Gene ID: (Human) 91833