Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ARELRVSCM QRKKVQIHSW DPSSLASDRF NRILANTNTD QLFTVNQVEA GGSKYGIITM RGLTTPELRV YPHKTLYVPN RKVNSMCWAS LNHLDSHLLL CFVGLADTPS CAVLLPASLF IGSFPGMRRP GMLCSFQIPD AWSCAWSLSI HAYHSFSTGL SQQVLLTNVV TGHQQSFGTS SDVLAQQFAI MTPLLFNGCR SGEIFGIDLR CGNQGSGWKA ICLSHDSAVT SLQILQDGQF LVSSDMTGTI KLWDLRATKC VTQYEGHVNN SAYLPVHVNE EEGVVAAVGQ DCYTRIWSLR HGHLLTTIPS PYPASENDIP SVAFSSRLGG FRGAPGLLMA VREDLYCFSY G (46-395 aa encoded by BC111778)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DDB1 and CUL4 associated factor 4-like 2; DDB1- and CUL4-associated factor 4-like protein 2; WD repeat domain 21C; WD repeat-containing protein 21C
Gene Aliases: DCAF4L2; WDR21C
UniProt ID: (Human) Q8NA75
Entrez Gene ID: (Human) 138009
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support