Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: FSISVLDIET RKIVREFSGH QGQINDMAFS PDGRWLISAA MDCSIRTWDL PSGCLIDCFL LDSAPLNVSM SPTGDFLATS HVDHLGIYLW SNISLYSVVS LRPLPADYVP SIVMLPGTCQ TQDVEVSEET VEPSDELIEY DSPEQLNEQL VTLSLLPESR WKNLLNLDVI KKKNKPKEPP KVPKSAPFFI PTIPGLVPRY AAPEQNNDPQ QSKVVNLGVL AQKSDFCLKL EEGLVNNKYD TALNLLKESG PSGIETELRS LSPDCGGSIE VMQSFLKMIG MMLDRKRDFE LAQAYLALFL KLHLKMLPSE PVLLEEITNL SSQVEENWTH LQSLFNQ
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Mutations in this gene have been associated with adult-onset primary open-angle glaucoma (POAG).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: T-cell activation WD repeat protein; T-cell activation WD repeat-containing protein; TA-WDRP; WD repeat-containing protein 36
Gene Aliases: 5730444A13Rik; GLC1G; TA-WDRP; TAWDRP; UTP21; WDR36
UniProt ID: (Human) Q8NI36
Entrez Gene ID: (Human) 134430, (Mouse) 225348
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support