Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLRWLIGGGR EPQGLAEKSP LQTIGEEQTQ NPYTELLVLK AHHDIVRFLV QLDDYRFASA GDDGIVVVWN AQTGEKLLEL NGHTQKITAI ITFPSLESCE EKNQLILTAS ADRTVIVWDG DTTRQVQRIS CFQSTVKCLT VLQRLDVWLS GGNDLCVWNR KLDLLCKTSH LSDTGISALV EIPKNCVVAA VGKELIIFRL VAPTEGSLEW DILEVKRLLD HQDNILSLIN VNDLSFVTGS HVGELIIWDA LDWTMQAYER NFWDPSPQLD TQQEIKLCQK SNDISIHHFT CDEENVFAAV GRGLYVYSLQ MKRVIACQKT AHDSNVLHVA RLPNRQLISC SEDGSVRIWE LREKQQLAAE PVPTGFFNMW GFGRVSKQAS QPVKKQQENA TSCSLELIGD LIGHSSSVEM FLYFEDHGLV TCSADHLIIL WKNGERESGL RSLRLFQKLE ENGDLYLAV
Non-catalytic component of the C9orf72-SMCR8 complex, a complex that has guanine nucleotide exchange factor (GEF) activity and regulates autophagy. The C9orf72-SMCR8 complex promotes the exchange of GDP to GTP, converting inactive GDP-bound RAB8A and RAB39B into their active GTP-bound form, thereby promoting autophagosome maturation. As part of the C9orf72-SMCR8 complex, stimulates RAB8A and RAB11A GTPase activity in vitro, however WDR42 is shown not be an essential complex component for this function. The C9orf72-SMCR8 complex also acts as a negative regulator of autophagy initiation by interacting with the ULK1/ATG1 kinase complex and inhibiting its protein kinase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: WD repeat-containing protein 41
Gene Aliases: MSTP048; WDR41
UniProt ID: (Human) Q9HAD4
Entrez Gene ID: (Human) 55255
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support