Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: GELEEKTDRM PQDELGQERR DLEPENREEG QERRVSDIQS KAGISRESLV
Sequence homology: Dog: 100%; Human: 100%; Pig: 85%
WDR66 contains 9 WD repeats. This protein encoded by WDR66 belongs to the WD repeat-containing family of proteins, which function in the formation of protein-protein complexes in a variety of biological pathways. This family member appears to function in the determination of mean platelet volume (MPV), and polymorphisms in this gene have been associated with variance in MPV. Alternative splicing of this gene results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 251; WD repeat-containing protein 66
Gene Aliases: CaM-IP4; CFAP251; WDR66
UniProt ID: (Human) Q8TBY9
Entrez Gene ID: (Human) 144406
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support