Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MDPGDDWLVE SLRLYQDFYA FDLSGATRVL EWIDDKGVFV AGYESLKKNE ILHLKLPLRL SVKENKGLFP ERDFKVRHGG FSDRSIFDLK HVPHTRLLVT SGLPGCYLQV WQVAEDSDVI KAVSTIAVHE KEESLWPRVA VFSTLAPGVL HGARLRSLQV VDLESRKTTY TSDVSDSEEL SSLQVLDADT FAFCCASGRL GLVDTRQKWA PLENRSPGPG SGGERWCAEV GSWGQGPGPS IASLGSDGRL CLLDPRDLCH PVSSVQCPVS VPSPDPELLR VTWAPGLKNC LAISGFDGTV QVYDATSWDG TRSQDGTRSQ VEPLFTHRGH IFLDGNGMDP APLVTTHTWH PCRPRTLLSA TNDASLHVWD WVDLCAPR
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ00296 protein; WD repeat-containing protein 73
Gene Aliases: GAMOS; HSPC264; WDR73
UniProt ID: (Human) Q6P4I2
Entrez Gene ID: (Human) 84942
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support