Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: NDSRPENPG PLGPISGVGG GGLGSGSDDN ALKQELPRSV HGLSGNWLAY WQYEIGVSQQ DAHFHFHQIR LQSFPGHSGA VKCVAPLSSE DFFLSGSKDR TVRLWPLYNY GDGTSETAPR LVYTQHRKSV FFVGQLEAPQ HVVSCDGAVH VWDPFTGKTL RTVEPLDSRV PLTAVAVMPA PHTSITMASS DSTLRFVDCR KPGLQHEFRL GGGLNPGLVR ALAISPSGRS VVAGFSSGFM VLLDTRTGLV LRGWPAHEGD ILQIKAVEGS VLVSSSSDHS LTVWKELEQK PTHHYKSASD PIHTFDLYGS EVVTGTVSNK IGVCSLLEPP SQATTKLSSE NFRGTLTSLA LLPTKRHLLL GSDNGVIRLL A
WD repeat-containing protein 81 (WDR81) is a 1941 amino acid protein, which contains one BEACH domain and five WD repeats. WDR81 is widely expressed in the brain. WDR81 has an association with cerebellar ataxia, mental retardation, and dysequilibrium syndrome 2. WDR81 exists as six isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protein phosphatase 1, regulatory subunit 166; WD repeat-containing protein 81
Gene Aliases: BC054822; CAMRQ2; Gm883; mFLJ00182; PPP1R166; SORF-2; WDR81
UniProt ID: (Human) Q562E7, (Mouse) Q5ND34
Entrez Gene ID: (Human) 124997, (Mouse) 192652
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support