Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: WSWLLFRSLQ RAPSWSFPSN LGTKTADLKG ASELPTPCHE CREDNDGEGH ARPQSGMKPL TEGMRKNGTW LQATAATTRD CGVNPEEADS A
Highest antigen sequence identity to the following orthologs: Mouse - 24%, Rat - 34%.
WDR85 is a WD repeat-containing protein that plays a role in the first step of diphthamide biosynthesis (Carette et al., 2009 [PubMed 19965467]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Diphthamide biosynthesis protein 7; Diphthine methyltransferase; RP11-48C7.3; WD repeat domain 85; WD repeat-containing protein 85
Gene Aliases: C9orf112; DPH7; RRT2; WDR85
UniProt ID: (Human) Q9BTV6
Entrez Gene ID: (Human) 92715
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support