Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive test controls include: RAW264.7, Mouse spleen.
Immunogen sequence: QLPSRSGVDS PRSGPRPPLP PDRPSAGAPP PPPPSTSIRN GFQDSPCEDE WESRFYFHPI SDLPPPEPYV QTTKSYPSKL ARNESRSGSN RRERGAPPLP P
This gene encodes a protein that plays an important role in the organization of the actin cytoskeleton. The encoded protein binds to a region of Wiskott-Aldrich syndrome protein that is frequently mutated in Wiskott-Aldrich syndrome, an X-linked recessive disorder. Impairment of the interaction between these two proteins may contribute to the disease. Two transcript variants encoding the same protein have been identified for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC111041; Protein PRPL-2; testicular tissue protein Li 226; WAS/WASL interacting protein family, member 1; WAS/WASL-interacting protein family member 1; WASP interacting protein; WASP-interacting protein; Wiskott-Aldrich syndrome protein interacting protein; Wiskott-Aldrich syndrome protein-interacting protein
Gene Aliases: AI115543; D2Ertd120e; PRPL-2; WAS2; WASPIP; WIP; WIPF1
UniProt ID: (Human) O43516, (Rat) Q6IN36, (Mouse) Q8K1I7
Entrez Gene ID: (Human) 7456, (Rat) 117538, (Mouse) 215280
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support