Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 13
Recommended positive controls: 293T, A431, HeLa, HepG2, mouse heart.
Predicted reactivity: Mouse (96%), Rat (96%), Xenopus laevis (81%), Rabbit (97%), Chicken (85%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Wnt-11; wingless-related MMTV integration site 11; wingless-type MMTV integration site family, member 11
Gene Aliases: HWNT11; Wnt-11; WNT11
UniProt ID: (Human) O96014, (Mouse) P48615
Entrez Gene ID: (Human) 7481, (Rat) 140584, (Mouse) 22411
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support