Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 5
It is not recommended to aliquot this antibody.
This antibody works to detect both isoforms of human Wnt5a protein.
Wnt5A is a secreted protein found in the extracellular region that belongs to the Wnt family. It is a proto-oncogene that functions in modulating cell fate and behavior during vertebrate development and has an important role in Wnt signaling pathway. It is a glycoprotein that plays an important role in cell communication and development. Wnt5A acts as a ligand for members of the frizzled family of seven transmembrane receptors. It is suspected that Wnt5A is a developmental protein. Wnt5A is a signaling molecule which affects the development of discrete regions of tissues and is likely to signal over only few cell diameters.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OTTHUMP00000212837; OTTHUMP00000212861; Protein Wnt-5a; wingless-related MMTV integration site 5A; wingless-type MMTV integration site 5A; wingless-type MMTV integration site family member 5A; wingless-type MMTV integration site family, member 5A; WNT-5A protein
Gene Aliases: 8030457G12Rik; hWNT5A; Wnt-5a; WNT5A
UniProt ID: (Human) P41221, (Mouse) P22725
Entrez Gene ID: (Human) 7474, (Rat) 64566, (Mouse) 22418
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support