Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Positive control: HeLa cell lysate, HepG2 cell lysate, SW480 cell lysate, Jurkat cell lysate, Raji cell lysate, NIH/3T3 cell lysate, mouse brain tissue lysate, mouse embryo tissue lysate, rat brain tissue lysate, NIH/3T3, HeLa.
Predicted band size: 38.8 kDa
Subcellular Location: Secreted, extracellular space, extracellular matrix.
WNT8A, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein Wnt-8a; Protein Wnt-8d; Stimulated by retinoic acid gene 11 protein; wingless-related MMTV integration site 8A; wingless-related MMTV integration site 8D; wingless-type MMTV integration site family member 8A; wingless-type MMTV integration site family, member 8A; wnt-8; WNT8d
Gene Aliases: Stra11; Wnt-8A; Wnt-8D; WNT8A; WNT8D
UniProt ID: (Human) Q9H1J5, (Mouse) Q64527
Entrez Gene ID: (Human) 7478, (Mouse) 20890, (Rat) 291678
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support