Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • WRAP53 Antibodies

            Proteintech

            WRAP53 Polyclonal Antibody

            12 References
            View all (14) WRAP53 antibodies

            Promotions

            View available promotion(s)

            Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # 14761-1-AP
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            492.00
            150 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite WRAP53 Polyclonal Antibody

            • Antibody Testing Data (25)
            Application
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 25

            WRAP53 Antibody (14761-1-AP) in IHC (P)

            Immunohistochemistry of paraffin-embedded mouse testis tissue slide using 14761-1-AP (WRAP53 antibody) at dilution of 1:200 (under 40x lens) heat mediated antigen retrieved with Tris-EDTA buffer (pH 9).
            View Product

            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            WRAP53 Antibody in Western Blot (WB)
            WRAP53 Antibody in Western Blot (WB)
            WRAP53 Antibody in Western Blot (WB)
            WRAP53 Antibody in Western Blot (WB)
            WRAP53 Antibody in Immunoprecipitation (IP)

            Product Details

            14761-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:2,000
            View 8 publications 8 publications

            Immunohistochemistry (IHC)

            -
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            1:50-1:500
            -

            Immunocytochemistry (ICC/IF)

            -
            View 3 publications 3 publications

            Immunoprecipitation (IP)

            0.5-4.0 µg
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            WRAP53 Fusion Protein Ag6522 (166-548 aa encoded by BC002336)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.2 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: TQPENFLKG CKWAPDGSCI LTNSADNILR IYNLPPELYH EGEQVEYAEM VPVLRMVEGD TIYDYCWYSL MSSAQPDTSY VASSSRENPI HIWDAFTGEL RASFRAYNHL DELTAAHSLC FSPDGSQLFC GFNRTVRVFS TARPGRDCEV RATFAKKQGQ SGIISCIAFS PAQPLYACGS YGRSLGLYAW DDGSPLALLG GHQGGITHLC FHPDGNRFFS GARKDAELLC WDLRQSGYPL WSLGREVTTN QRIYFDLDPT GQFLVSGSTS GAVSVWDTDG PGNDGKPEPV LSFLPQKDCT NGVSLHPSLP LLATASGQRV FPEPTESGDE GEELGLPLLS TRHVHLECRL QLWWCGGAPD SSIPDDHQGE KGQGGTEGGV GELI (166-548 aa encoded by B C002336)

            Target Information

            TXNDC1 is a thioredoxin -related protein with disulfide reductase activity.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: Telomerase Cajal body protein 1; WD repeat containing, antisense to TP53; WD repeat domain 79; WD repeat-containing protein 79; WD repeat-containing protein WRAP53; WD40 protein Wrap53; WD40 repeat-containing protein antisense to TP53; WD40 repeat-containing protein antisense to TP53 gene; WD40 repeat-containing protein antisense to TP53 gene homolog; WD40 repeat-containing protein encoded by RNA antisense to p53; WD40 repeat-containing protein encoding RNA antisense to p53; WRAP53beta

            View more View less

            Gene Aliases: BC021790; DKCB3; TCAB1; WDR79; WRAP53

            View more View less

            UniProt ID: (Human) Q9BUR4, (Rat) Q5XII5, (Mouse) Q8VC51

            View more View less

            Entrez Gene ID: (Human) 55135, (Rat) 287432, (Mouse) 216853

            View more View less

            Function(s)

            RNA binding protein binding protein complex binding chaperone binding telomerase RNA binding

            Process(es)

            telomere formation via telomerase positive regulation of telomerase activity telomere maintenance via telomerase positive regulation of telomere maintenance via telomerase establishment of protein localization to telomere scaRNA localization to Cajal body telomerase RNA localization to Cajal body positive regulation of establishment of protein localization to telomere protein localization to Cajal body
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-qdb8m:80/100.66.73.94:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0