Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ARRENILVMR LATKEQEMQE CTTQIQYLKQ VQQPSVAQLR STMVDPAINL FFLKMKGELE QTKDKLEQAQ NELSAWKFTP DR
Wilms' tumor (WT) is an embryonal malignancy of the kidney that affects 1 in 10,000 infants and is observed in both sporadic and inherited forms. The Wilms' tumor protein (WT1) binds the DNA sequence GCGGGGGCG, a recognition element common to the early growth response (Egr) family of Zn2+ finger transcriptional activators, and functions as a transcriptional repressor. WTAP (Wilms tumor 1-associating protein) is a ubiquitously expressed nuclear protein that interacts with WT1 and may be involved in regulating mRNA splicing. WTAP is found in nuclear speckles, where it regulates the G2/M cell cycle transition by binding to the 3' UTR of cyclin A2, thus enhancing its stability. Additionally, WTAP inhibits expression of WT1 target genes and is able to impair the ability of WT1 to bind DNA. Two isoforms of WTAP exist due to alternative splicing events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Female-lethal(2)D homolog; hFL(2)D; PNAS-132; Pre-mRNA-splicing regulator WTAP; putative pre-mRNA splicing regulator female-lethal(2D); Wilms tumor 1-associating protein; Wilms' tumour 1-associating protein; WT1-associated protein
Gene Aliases: KIAA0105; Mum2; WTAP
UniProt ID: (Human) Q15007
Entrez Gene ID: (Human) 9589
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support