Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse and Rat
A novel protein designated XAB2 (XPA-binding protein 2) interacts with XPA, a factor central to both nucleotide excision repair subpathways. Data suggests that XAB2 is a novel component involved in transcription-coupled repair and transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: crn-related protein kim1; DKFZp762C1015; HCNP protein; Pre-mRNA-splicing factor SYF1; Protein HCNP; SYF1 homolog, RNA splicing factor; SYF1 pre-mRNA-splicing factor; XPA-binding protein 2
Gene Aliases: HCNP; HCRN; KIAA1177; NTC90; PP3898; SYF1; XAB2
UniProt ID: (Human) Q9HCS7
Entrez Gene ID: (Human) 56949
Molecular Function: RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support