Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: HeLa, NIH-3T3, JC, BCL-1.
Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (81%), Bovine (100%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
A novel protein designated XAB2 (XPA-binding protein 2) interacts with XPA, a factor central to both nucleotide excision repair subpathways. Data suggests that XAB2 is a novel component involved in transcription-coupled repair and transcription.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: crn-related protein kim1; DKFZp762C1015; HCNP protein; Pre-mRNA-splicing factor SYF1; Protein HCNP; SYF1 homolog, RNA splicing factor; SYF1 pre-mRNA-splicing factor; XPA-binding protein 2
Gene Aliases: 0610041O14Rik; AV025587; HCNP; HCRN; KIAA1177; NTC90; PP3898; SYF1; XAB2
UniProt ID: (Human) Q9HCS7, (Mouse) Q9DCD2
Entrez Gene ID: (Human) 56949, (Mouse) 67439
Molecular Function: RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support