Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MVVMARLSRP ERPDLVFEEE DLPYEEEIMR NQFSVKCWLR YIEFKQGAPK PRLNQLYERA LKLLPCSYKL WYRYLKARRA QVKHRCVTDP AYEDVNNCHE RAFVFMHKMP RLWLDYCQFL MDQGRVTHTR RTFDRALRAL PITQHSRIWP LYLRFLRSHP LPETAVRGYR RFLKLSPESA EEYIEYLKSS DRLDEAAQRL ATVVNDERFV SKAGKSNYQL WHELCDLISQ NPDKVQSLNV DAIIRGGLTR FTDQLGKLWC SLADYYIRSG HFEKARDVYE EAIRTVMTVR DFTQVFDSYA QFEESMIAAK METASELGRE EEDDVDLELR LARFEQLISR RPLLLNSVLL RQNPHHVHEW HKRVALHQGR PREIINTYTE AVQTVDPFKA TGKPHTLWVA FAKFYEDNGQ LDDARVILEK ATKVNFKQVD DLASVWCQCG ELELRHENYD EALRLLRKAT ALPARRAEYF DGSEPVQNRV YKSLKVWSML ADLEESLGTF QSTKAVYDRI LDLRIATPQI VINYAMFLEE HKYFEESFKA YERGISLFKW PNVSDIWSTY LTKFIARYGG RKLERARDLF EQALDGCPPK YAKTLYLLYA QLEEEWGLAR HAMAVYERAT RAVEPAQQYD MFNIYIKRAA EIYGVTHTRG IYQKAIEVLS DEHAREMCLR FADMECKLGE IDRARAIYSF CSQICDPRTT GAFWQTWKDF EVRHGNEDTI KEMLRIRRSV QATYNTQVNF MASQMLKVSG SATGTVSDLA PGQSGMDDMK LLEQRAEQLA AEAERDQPLR AQSKILFVRS DASREELAEL AQQVNPEEIQ LGEDEDEDEM DLEPNEVRLE QQSVPAAVFG SLKED
A novel protein designated XAB2 (XPA-binding protein 2) interacts with XPA, a factor central to both nucleotide excision repair subpathways. Data suggests that XAB2 is a novel component involved in transcription-coupled repair and transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: crn-related protein kim1; DKFZp762C1015; HCNP protein; Pre-mRNA-splicing factor SYF1; Protein HCNP; SYF1 homolog, RNA splicing factor; SYF1 pre-mRNA-splicing factor; XPA-binding protein 2
Gene Aliases: HCNP; HCRN; KIAA1177; NTC90; PP3898; SYF1; XAB2
UniProt ID: (Human) Q9HCS7
Entrez Gene ID: (Human) 56949
Molecular Function: RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support