Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: RNPTPDQKRE DDQGAAEIQV PDLEADLQEL CQTKTGDGCE GGTDVKGKIL PKAEHFKMPE AGEGKSQV
This gene is a member of the XAGE subfamily, which belongs to the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in normal testis, and in Ewing's sarcoma, rhabdomyosarcoma, a breast cancer and a germ cell tumor. The protein encoded by this gene shares a sequence similarity with other GAGE/PAGE proteins. Because of the expression pattern and the sequence similarity, this protein also belongs to a family of CT (cancer-testis) antigens.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 12.2; cancer/testis antigen family 12, member 2; CT12.2; G antigen family D member 3; X antigen family member 2; X antigen family, member 2; X antigen family, member 2B; XAGE-2
Gene Aliases: CT12.2; GAGED3; XAGE-2; XAGE2; XAGE2B
UniProt ID: (Human) Q96GT9
Entrez Gene ID: (Human) 9502
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support