Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPPKVTSELL RQLRQAMRNS EYVTEPIQAY IIPSGDAHQS EYIAPCDCRR AFVSGFDGSA GTAIITEEHA AMWTDGRYFL QAAKQMDSNW TLMKMGLKDT PTQEDWLVSV LPEGSRVGVD PLIIPTDYWK KMAKVLRSAG HHLIPVKENL VDKIWTDRPE RPCKPLLTLG LDYTGISWKD KVADLRLKMA ERNVMWFVVT ALDEIAWLFN LRGSDVEHNP VFFSYAIIGL ETIMLFIDGD RIDAPSVKEH LLLDLGLEAE YRIQVHPYKS ILSELKALCA DLSPREKVWV SDKASYAVSE TIPKDHRCCM PYTPICIAKA VKNSAESEGM RRAHIKDAVA LCELFNWLEK EVPKGGVTEI SAADKAEEFR RQQADFVDLS FPTISSTGPN GAIIHYAPVP ETNRTLSLDE VYLIDSGAQY KDGTTDVTRT MHFGTPTAYE KECFTYVLKG HIAVSAAVFP TGTKGHLLDS FARSALWDSG LDYLHGTGHG VGSFLNVHEG PCGISYKTFS DEPLEAGMIV TDEPGYYEDG AFGIRIENVV LVVPVKTKYN FNNRGSLTFE PLTLVPIQTK MIDVDSLTDK ECDWLNNYHL TCRDVIGKEL QKQGRQEALE WLIRETQPIS KQH
This gene encodes the cytosolic form of a metalloaminopeptidase that catalyzes the cleavage of the N-terminal amino acid adjacent to a proline residue. The gene product may play a role in degradation and maturation of tachykinins, neuropeptides, and peptide hormones. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aminoacylproline aminopeptidase; aminopeptidase P, cytosolic; Cytosolic aminopeptidase P; OTTHUMP00000020457; OTTHUMP00000058856; sAmp; Soluble aminopeptidase P; X-Pro aminopeptidase 1; X-prolyl aminopeptidase (aminopeptidase P) 1, soluble; X-prolyl aminopeptidase 1, soluble; Xaa-Pro aminopeptidase 1
Gene Aliases: APP1; SAMP; XPNPEP; XPNPEP1; XPNPEPL; XPNPEPL1
UniProt ID: (Human) Q9NQW7
Entrez Gene ID: (Human) 7511
Molecular Function: protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support