Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Mouse, Hamster, Rabbit (100%) Rat, Cat, Bovine, Dog, Elephant, Horse, Pig (94%).
Xenotropic and polytropic retrovirus receptor (XPR1) is an Orphan-U GPCR with an unknown ligand. ESTs have been isolated from brain, breast, kidney, pancreas, placenta, prostate, skeletal muscle, testis, and tonsil libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Phosphate exporter SLC53A1; Protein SYG1 homolog; receptor for MCF virus 1; Rmc-1; RP11-46A10.1; Solute carrier family 53 member 1; suppressor of yeast G deletion; susceptibility to xenotropic virus; X-receptor; xenotropic and polytropic murine leukemia virus receptor; Xenotropic and polytropic murine leukemia virus receptor X3; xenotropic and polytropic murine retrovirus receptor; Xenotropic and polytropic retrovirus receptor 1; Xenotropic and polytropic retrovirus receptor 1 homolog
Gene Aliases: IBGC6; Rmc-1; Rmc1; SLC53A1; Sxv; SYG1; X3; XPR1; XR
UniProt ID: (Human) Q9UBH6, (Mouse) Q9Z0U0, (Chinese hamster) Q9QZ70
Entrez Gene ID: (Rabbit) 100339244, (Human) 9213, (Mouse) 19775, (Chinese hamster) 100689082
Molecular Function: secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support