Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Immunogen sequence: TAPVPARAQ GAVTGKPRGE GTEPRRPRAG PEELGKILQG VVVVLSGFQN PFRSELRDKA LELGAKYRPD WTRDSTHLIC AFANTPKYSQ VLGLGGRIVR KEWVLDCHRM RRRLPSQRYL MAGPGSSSEE DEASHSGGSG DEAPKLPQKQ PQTKTKPTQA AGPSSPQKPP TPEETKAASP VLQEDIDIEG VQSEGQDNGA EDSGDTEDEL RRVAEQKEHR LPPGQEENGE DPYAGSTDEN TDSEEHQEPP DLPVPELPDF FQGKHFFLYG EFPGDERRKL IRYVTAFNGE LEDYMSDRVQ FVITAQEWDP SFEEALMDNP SLAFVRPRWI YSCNEKQKLL PHQLYGVVPQ A (284-633 aa encoded by BC023593)
XRCC1 (X-ray cross complementing factor-1) is involved in DNA base excision repair. XRCC1 is a 70 kDa protein that has been shown to be physically associated with other DNA repair enzymes, including poly (ADP-ribose) polymerase (PARP), DNA Ligase III and DNA polymerase beta. XRCC1 contains a BRCT domain (for BRCA1 carboxyl terminus) which was originally found in the tumor suppressor protein BRCA1. Recently, the BRCT domain has been shown to mediate the protein-protein interaction of XRCC1 and DNA ligase III-alpha.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA repair protein XRCC1; X-ray repair complementing defective repair in Chinese hamster cells 1; X-ray repair cross-complementing protein 1
Gene Aliases: RCC; Xrcc-1; XRCC1
UniProt ID: (Human) P18887, (Mouse) Q60596
Entrez Gene ID: (Human) 7515, (Mouse) 22594
Molecular Function:
DNA metabolism protein
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support