Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MCSAFHRAE SGTELLARLE GRSSLKEIEP NLFADEDSPV HGDILEFHGP EGTGKTEMLY HLTARCILPK SEGGLEVEVL FIDTDYHFDM LRLVTILEHR LSQSSEEIIK YCLGRFFLVY CSSSTHLLLT LYSLESMFCS HPSLCLLILD SLSAFYWIDR VNGGESVNLQ ESTLRKCSQC LEKLVNDYRL VLFATTQTIM QKASSSSEEP SHASRRLCDV DIDYRPYLCK AWQQLVKHRM FFSKQDDSQS SNQFSLVSRC LKSNSLKKHF FIIGESGVEF C (1-280 aa encoded by BC042137)
The x-ray repair cross-complementing (XRCC) proteins are responsible for efficiently repairing and maintaining genetic stability following DNA base damage. These genes share sequence similarity with the yeast DNA repair protein Rad5. XRCC2 and XRCC3 are both involved in maintaining chromosome stability during cell division. XRCC2 is required for efficient repair of DNA double-strand breaks by homologous recombination between sister chromatids, and XRCC3 interacts directly with Rad51 to cooperate with Rad51 during recombinational repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781P0919; DNA repair protein XRCC2; recA/RAD51 family protein; X-ray repair complementing defective repair in Chinese hamster cells 2; X-ray repair cross-complementing protein 2
Gene Aliases: 4921524O04Rik; 8030409M04Rik; FANCU; RAD51; RecA; XRCC2
UniProt ID: (Human) O43543, (Mouse) Q9CX47
Entrez Gene ID: (Human) 7516, (Mouse) 57434
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support