Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MAGAPDERRR GPAAGEQLQQ QHVSCQVFPE RLAQGNPQQG FFSSFFTSNQ KCQLRLLKTL ETNPYVKLLL DAMKHSGCAV NKDRHFSCED CNGNVSGGFD ASTSQIVLCQ NNIHNQAHMN RVVTHELIHA FDHCRAHVDW FTNIRHLACS EVRAANLSGD CSLVNEIFRL HFGLKQHHQT CVRDRATLSI LAVRNISKEV AKKAVDEVFE SCFNDHEPFG RIPHNKTYAR YAHRDFENRD RYYSNI
The protein encoded by this gene is amplified in glioblastomas and interacts with the DNA binding subunit of DNA-dependent protein kinase. This kinase is involved in double-strand break repair (DSB), and higher expression of the encoded protein increases the efficiency of DSB. In addition, comparison to orthologous proteins strongly suggests that this protein is a metalloprotease important in the biosynthesis of mitochondrial ATPase. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ku70 binding protein 3; Ku70-binding protein 3; Mitochondrial inner membrane protease ATP23 homolog; XRCC6 binding protein 1; XRCC6-binding protein 1
Gene Aliases: ATP23; KUB3; XRCC6BP1
UniProt ID: (Human) Q9Y6H3
Entrez Gene ID: (Human) 91419
Molecular Function: metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support