Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockout to ensure that the antibody binds to the antigen stated.
Immunogen sequence: PTAQHLRQSS FEIPDDVPLP AGWEMAKTSS GQRYFLNHID QTTTWQDPRK AMLSQMNVTA PTSPPVQQNM MNSASGPLPD GWEQAMTQDG EIYYINHKNK TTSWLDPRLD PRFAMNQRIS QSAPVKQPPP LAPQSPQGGV MGGSNSNQQQ QMRLQQLQME KERLRLKQQE LLRQAMRNIN PSTANSPKCQ ELALRSQLPT LEQDGGTQNP VSSPGMSQEL RTMTTNSSDP FLNSGTYHSR DESTDSGLSM SSYSVPRTPD DFLNSVDEMD TGDTINQSTL PSQQNRFPDY LEAIPGTNVD LGTLEGDGMN IEGEELMPSL QEALSSDILN DMESVLAATK LDKESFLTWL
YAP (Yes-Associated Protein) is a founding member of the WW domain family of proteins, transcriptional co-activator and oncogene. Its gene is transcriptionally activated in squamous cell carcinoma cells. YAP can act both as a coactivator and corepressor and is the critical downstream regulatory target in the Hippo signaling pathway. The Hippo signaling pathway plays a pivotal role in organ size control and tumour suppression by restricting proliferation and promoting apoptosis. Also plays key roles in tissue tension, 3D tissue shape, cell proliferation, cell death, and cell migration. Mutations in the IRF5 gene results in coloboma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 65 kDa Yes-associated protein; Protein yorkie homolog; Transcriptional coactivator YAP1; Yes-associated protein 1; yes-associated protein 2; Yes-associated protein YAP65 homolog; yes-associated protein, 65 kDa; yorkie homolog
Gene Aliases: AI325207; COB1; YAP; YAP1; YAP2; YAP65; YKI; Yorkie
UniProt ID: (Human) P46937, (Mouse) P46938
Entrez Gene ID: (Human) 10413, (Rat) 363014, (Mouse) 22601
Molecular Function:
transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support