Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: QIVHVRGDSE TDLEALFNAV MNPKTANVPQ TVPMRLRKLP DSFFKPPEPK SHSRQASTDA GTAGALTPQH VRAHSSPASL QLGAVSPGTL TPTGVVSGPA ATPTAQHLR
YAP (Yes-Associated Protein) is a founding member of the WW domain family of proteins, transcriptional co-activator and oncogene. Its gene is transcriptionally activated in squamous cell carcinoma cells. YAP can act both as a coactivator and corepressor and is the critical downstream regulatory target in the Hippo signaling pathway. The Hippo signaling pathway plays a pivotal role in organ size control and tumour suppression by restricting proliferation and promoting apoptosis. Also plays key roles in tissue tension, 3D tissue shape, cell proliferation, cell death, and cell migration. Mutations in the IRF5 gene results in coloboma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 65 kDa Yes-associated protein; Protein yorkie homolog; Transcriptional coactivator YAP1; Yes-associated protein 1; yes-associated protein 2; Yes-associated protein YAP65 homolog; yorkie homolog
Gene Aliases: COB1; YAP; YAP1; YAP2; YAP65; YKI
UniProt ID: (Human) P46937
Entrez Gene ID: (Human) 10413
Molecular Function:
transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support