Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EREALETER VRANARALRL GLEALAANHQ QLFTDGRSWG SFTVLDNSAW YQKQHLVDFL AAVGGHFRMG TLLSRQSVQL RLKSPEGMSL AEFFYQVLQA YDFYYLFQRY GCRVQLGGSD QLGNIMSGYE FINKLTGEDV FGITVPLITS TTGAKLGKSA GNAVWLNRDK TSPFELYQFF VRQPDDSVER YLKLFTFLPL PEIDHIMQLH VKEPERRGPQ KRLAAEVTKL VHGREGLDSA KRCTQALYHS SIDALEVMSD QELKELFKEA PFSEFFLDPG TSVLDTCRKA NAIPDGPRGY RMITEGGVSI NHQQVTNPES VLIVGQHILK NGLSLLKIGK RNFYIIKWLQ L (128-477 aa encoded by BC015625)
This gene encodes a mitochondrial protein that catalyzes the attachment of tyrosine to tRNA(Tyr). Mutations in this gene are associated with myopathy with lactic acidosis and sideroblastic anemia type 2 (MLASA2).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 6.1.1.1; tyrosine tRNA ligase 2, mitochondrial; Tyrosine--tRNA ligase, mitochondrial; tyrosyl trna synthetase; Tyrosyl-tRNA synthetase; tyrosyl-tRNA synthetase 2, mitochondrial; tyrosyl-tRNA synthetase, mitochondrial; TyrRS
Gene Aliases: 2210023C10Rik; CGI-04; MLASA2; MT-TYRRS; RGD1311696; TYRRS; YARS2
UniProt ID: (Human) Q9Y2Z4, (Mouse) Q8BYL4, (Rat) Q5I0L3
Entrez Gene ID: (Human) 51067, (Mouse) 70120, (Rat) 287924
Molecular Function: aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support