Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAAPILRSFS WGRWSGTLNL SVLLPLGLRK AHSGAQGLLA AQKARGLFKD FFPETGTKIE LPELFDRGTA SFPQTIYCGF DPTADSLHVG HLLALLGLFH LQRAGHNVIA LVGGATARLG DPSGRTKERE ALETERVRAN ARALRLGLEA LAANHQQLFT DGRSWGSFTV LDNSAWYQKQ HLVDFLAAVG GHFRMGTLLS RQSVQLRLKS PEGMSLAEFF YQVLQAYDFY YLFQRYGCRV QLGGSDQLGN IMSGYEFINK LTGEDVFGIT VPLITSTTGA KLGKSAGNAV WLNRDKTSPF ELYQFFVRQP DDSVERYLKL FTFLPLPEID HIMQLHVKEP ERRGPQKRLA AEVTKLVHGR EGLDSAKRCT QALYHSSIDA LEVMSDQELK ELFKEAPFSE FFLDPGTSVL DTCRKANAIP DGPRGYRMIT EGGVSINHQQ VTNPESVLIV GQHILKNGLS LLKIGKRNFY IIKWLQL
This gene encodes a mitochondrial protein that catalyzes the attachment of tyrosine to tRNA(Tyr). Mutations in this gene are associated with myopathy with lactic acidosis and sideroblastic anemia type 2 (MLASA2).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 6.1.1.1; tyrosine tRNA ligase 2, mitochondrial; Tyrosine--tRNA ligase, mitochondrial; tyrosyl trna synthetase; Tyrosyl-tRNA synthetase; tyrosyl-tRNA synthetase 2, mitochondrial; TyrRS
Gene Aliases: CGI-04; MLASA2; MT-TYRRS; TYRRS; YARS2
UniProt ID: (Human) Q9Y2Z4
Entrez Gene ID: (Human) 51067
Molecular Function: aminoacyl-tRNA synthetase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support