Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: VQTILMPVN KVVQSFSTSK PPAILPVAAP TPVVPSSAPA AVAKVKTEPE TPGPSCLSQE GQTAVKTEES SELGNYVIKI DHLETIQQLL TAVVKKIPLI TAKSEDASCF SAKSVEQYYG WNIGKRRAAE WQRAMTMRKV LQEILEKNPR FHHLTPLKTK HIAHWCRCHG YTPPDPESLR NDGDSIEDVL TQIDSEPECP SSFSSADNLC RKLEDLQQFQ KREPENEEEV DILSLSEPVK INIKKEQEEK QEEVKFYLPP TPGSEFIGDV TQKIGITLQP VALHRNVYAS VVEDMILKAT EQLVNDILRQ ALAVGYQTAS HNRIPKEITV SNIHQAICNI PFLDFLTNKH MGILNEDQ (1066-1422 aa encoded by BC150273)
The YEATS domain-containing protein 2 (YEATS2) is a nuclear protein that is phosphorylated upon DNA damage by ATM or ATR and contains 1 YEATS domain. The gene encoding YEATS2 resides on chromosome 3, which contains about 214 million bases encoding over 1, 100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumor suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: YEATS domain-containing protein 2; YETS2
Gene Aliases: KIAA1197; YEATS2
UniProt ID: (Human) Q9ULM3
Entrez Gene ID: (Human) 55689
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support